Flame-retardant rain jacket, high visibility, Hedland 7223
Leverancier: SIOEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7223EF7278/1XL 7223EF7279/L 7223EF7279/3XL 7223EF7279/2XL 7223EF7279/1XL 7223EF7278/S 7223EF7278/M 7223EF7278/L 7223EF7278/2XL 7223EF7278/3XL 7223EF7279/M 7223EF7279/S
113-7229EA 216 EUR
113-7229 113-7238 113-7237 113-7236 113-7235 113-7234 113-7233 113-7232 113-7230 113-7231 113-7239 113-7240
Flame-retardant rain jacket, high visibility, Hedland 7223
Werkkleding
This waterproof and windproof jacket is made of anti-static Siopor® FR AST fabric: 100% polyester fabric with 100% FR PU coating + AST. Inside it features a fixed flame-retardant cotton lining with two pockets. The outer fabric is water-repellent, while the coating on the inside attracts moisture and keeps the skin dry.
  • High tear resistance, taped seams
  • Straight collar, provided for hood (hood available separately)
  • Zip closure under flap with touch and close fastening
  • One patched chest pocket, two patched pockets with gusset, two loops for badges, Napoleon pocket
  • Round inset sleeves, sleeve narrowing by press studs
  • Drawstring in hem
Back length: 85 cm
Weight: 240 g/m²
Certificaten: EN 531 (A B2 C1), EN 13034 (Type 6), EN 14116 (inside index 3; outside index 1), EN 1149-5, EN 343 (3-3), EN 471 (3-2)