High visibility rain jacket with arc protection, Nash 7227
Leverancier: SIOEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7227LK2143/S 7227LK2143/M 7227LK2143/L 7227LK2143/3XL 7227LK2143/2XL 7227LK2143/1XL
113-7247EA 447 EUR
113-7247 113-7246 113-7245 113-7244 113-7243 113-7242
High visibility rain jacket with arc protection, Nash 7227
Werkkleding
This high performance jacket is made of a 2-layer laminated fabric consisting of 60% FR Modacryl and 40% cotton (& AST) ripstop with PU laminate. The jacket features a fixed flame-retardant cotton lining but other fleeces and lining can be zipped in at any time.
  • Highly breathable, taped seams
  • Straight collar, provided for hood (hood available separately)
  • Zip closure under flap with touch and close fastening
  • One patched chest pocket, two patched pockets, two loops for badges, Napoleon pocket
  • Round inset sleeves, narrowing by touch and close fastening
  • Elasticated waist in the back
Back length: 85 cm
Weight: 310 g/m²
Certificaten: IEC 61482-2 (class 1), EN 11612 (A1 B1 C2 E1 F2 W33), EN 13034 (Type PB/6), EN 14116 (index 3), EN 11611 (class 2 - A1), EN 1149-5, EN 343 (3-3), EN 471 (2-2 until size S, 3-2 from size M on)