Flame-retardant rain trousers, high visibility, Kamber 7235
Leverancier: Sioen
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Siopor® Excell
113-7266EA 173 EUR
113-7266 113-7265 113-7268 113-7267 113-7262 113-7261 113-7264 113-7263 113-7271 113-7260 113-7270 113-7269
Flame-retardant rain trousers, high visibility, Kamber 7235
Werkkleding
Lightweight, high performance trousers made of Siopor Excell, a 3-layer fabric consisting of Dobby Ripstop polyester fabric + AST, a breathable FR PU coating, and an inherent FR knitting. The material is water-repellent on the outside and features a moisture-attracting coating on the inside.
  • Waterproof and windproof, highly breathable, anti-static
  • Taped seams
  • Supple, high tear resistance
  • Legs adjustable by press studs
  • Elasticated waist
Weight: 270 g/m²
Certificaten: EN 343 (3-3), EN 1149-5, EN 13034 (PB/6), EN ISO 11612 (A1 B2 C1 E1 F1 W33), EN ISO 14116, EN ISO 11611 (class 1 - A1A2)