His MultiTrap™ FF and HP filter plates prepacked with Sepharose™
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
28-4009-90 28-4009-89
28-4009-90EA 952 EUR
28-4009-90 28-4009-89
His MultiTrap™ FF and HP filter plates prepacked with Sepharose™
Microplaten Filtreerplaten
His MultiTrap™ FF and His MultiTrap™ HP are prepacked disposable 96-well plates for reproducible high throughput parallel purification of histidine-tagged recombinant proteins by Immobilised Metal ion Affinity Chromatography (IMAC).
  • Prepacked multiwell filter plates for high reproducibility in expression screening and small-scale purification of histidine-tagged proteins
  • Increased convenience and consistency with prepacked 96-well plates
  • No filtration needed - load unclarified sample directly, increase reproducibility, and save time
  • Use His MultiTrap™ 96-