Electrical and valve lockout kits
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
806187 806191 806184 805909 806181 806188 806186 806180 806182
BRDY806187EA 541.81 EUR
BRDY806187 BRDY806191 BRDY806184 238-0037 BRDY806181 BRDY806188 BRDY806186 BRDY806180 BRDY806182
Electrical and valve lockout kits
Veiligheidssloten
These kits include everything that is needed to lockout the most common mechanical and electrical control points.
Small adjustable lockout kit (supplied in nylon lockout pouch):
Adjustable gate valve device, APCLO with sheathed metal cable
Two Brady safety padlocks (red)
Two 25 mm safety lockouts (red)
Small universal valve lockout base clamping unit, small blocking arm
'Do Not Operate' tag and pen.
Valve lockout kit (supplied in cordura nylon satchel):
Large universal valve lockout base clamping unit, large blocking arm
Cable attachment, adjustable gate valve device
APLCO with sheathed metal cable
1 pack of safety padlocks (red)
1 pack of 25 mm safety lockouts (red)
'Do not operate' tag.
Gate valve lockout kit (supplied in satchel):
25 to 64 mm red gate valve lockout
64 to 127 mm red gate valve lockout
127 to 165 mm red gate valve lockout
165 to 254 mm red gate valve lockout
254 to 320 mm red gate valve lockout
1 pack of safety lockouts (red)
'Do not operate' tag.
Electrical lockout kit (supplied in nylon lockout pouch):
Universal multipole breaker, no-hole circuit breaker, no-hole circuit breaker (480 to 600 V)
277 V lockout cleat, 480 to 600 V lockout cleat
Miniature circuit breaker kit,
Two Brady safety padlocks (red)
Nylon non conductive hasp, red non conductive hasp
'Do Not Operate Electrician at work' tag and pen.
Valve and electrical lockout kit (supplied in polyester duffel bag):
Large universal valve lockout base clamping unit, large blocking arm
Cable attachment, adjustable gate valve device
APLCO with sheathed metal cable, large plug lockout device
227 V no-hole circuit breaker, 480 to 600 V no-hole circuit breaker, 480 to 600 V breaker block kit
Miniature circuit breaker demo kit (1 off each type)
Nylon lockout hasp
1 pack of safety padlocks (red)
1 pack of 25 mm safety lockouts (red)
'Do not operate' tag and pen.