Cell culture reagents, microcarriers, Cytodex 3
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17-0485-01 17-0485-03 17-0485-02
17-0485-01EA 381 EUR
17-0485-01 17-0485-03 17-0485-02
Cell culture reagents, microcarriers, Cytodex 3
Parels Microdragers
Cytodex 3, formed by chemically coupling a thin layer of denatured collagen to the cross-linked dextran matrix, is the microcarrier of choice for cells that may be difficult to culture in vitro, culture of primary cells or those with an epithelioid morphology, especially cells with low plating-efficiency or differentiated or sensitive cell-types which are normally difficult to grow, e.g. hepatocytes, endocrine cells.
  • Contains a layer of acid-denatured porcine collagen (gelatin) on the surface of the microporous matrix
  • Designed to yield harvested cells of