Cell culture reagents, microcarriers, Cytopore 1
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17-0911-01EA 1070 EUR
17-0911-01
Cell culture reagents, microcarriers, Cytopore 1
Parels Microdragers
Cytopore1 macroporous microcarriers are principally designed for use in suspension culture systems for growth of adherent recombinant Chinese Hamster Ovary (CHO) cells and the production of recombinant proteins.
  • Macroporous structure of Cytopore microcarriers encourages cell growth into the bead, while the micropores provide maximum nutrient availability
  • The characteristics enable growth of cells which require high recirculation rates and high nutrient-availability
  • Optimized for growth of r-CHO in stirred tank cultures but is applicable to cell lines requiring similar surface charge
  • Transparent for easy microscopic examination of attached cells