Cell culture reagents, microcarriers, Cytodex 1
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17-0448-01 17-0448-02 17-0448-03
17-0448-01EA 461 EUR
17-0448-01 17-0448-02 17-0448-03
Cell culture reagents, microcarriers, Cytodex 1
Parels Microdragers
Cytodex 1 is particularly suitable for most established cell lines and for production of viruses or cell products from cultures of primary cells and normal diploid cell strains.
  • General-purpose microcarrier for most established, anchorage-dependent cell lines
  • Contains positive charges throughout the microporous matrix
  • Transparent for easy microscopic examination of attached cells