Hygen™ Quick-Connect handles and frames
Leverancier: Rubbermaid Commercial Products
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FGQ58000 FGQ57000 FGQ59000
129-0756EA 78.8 EUR
129-0756 129-0755 129-0757
Hygen™ Quick-Connect handles and frames
Moppen
The Quick-Connect mechanism allows easy, time saving frame exchange.
  • Available in straight or ergonomic bend styles, in adjustable length from 115 to 186 cm
  • Extension poles make quick work of high reach cleaning tasks with length to 548,6 cm
  • Flat profile slides easily under furniture and equipment
  • Lightweight aluminium construction for improved productivity
  • One-step connecting mechanism speeds mop assembly and changeover
  • Unique pull type squeegee for water and debris collection
  • Replaceable hook-and-loop strips' cost efficiently extend product life