HPLC columns, Acclaim™ 120 C18
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
068981 068982 069689 069695 070077 071399 071400 071604 071605 071981 074812 075697 076186 076187 063691 066272 066273 059128 059129 059130 059131 059132 059133 059142 059143 059144 059145 059146 059147 059148 059149
555-7125EA 871 EUR
555-7125 555-7124 555-7116 555-7129 554-4459 555-7212 555-7214 555-7218 555-7219 555-7180 555-7507 555-7469 554-4463 554-4464 554-4452 554-4453 554-4454 554-4438 554-4439 554-4440 554-4441 554-4442 554-4443 554-4444 554-4445 554-4446 554-4447 554-4448 554-4449 554-4450 554-4451
HPLC columns, Acclaim™ 120 C18
Kolommen voor chromatografie
High density, monolayer C18 reversed phase columns for very good resolution in a variety of applications.
  • Very high surface coverage and very low metal content
  • Reproducible column-to-column performance
  • Extremely low bleed, fully compatible with MS
  • Stable between pH 2 and 8