Sterility culture media
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
STBMFTM12 STBMRFA34 STBMTSB12 STBMRFA12
MILXSTBMFTM12EA 139 EUR
MILXSTBMFTM12 MILXSTBMRFA34 MILXSTBMTSB12 MILXSTBMRFA12
Sterility culture media
Media voor microbiologie Geprepareerde media voor microbiologie
For sterility testing (fi ltration and direct inoculation) and bioburden testing. In the screw cap version, the rimless cap design minimizes the risks of cross contamination and optimizes the disinfection procedures, avoiding the risk of inhibition from disinfectant residuals.
  • Easy to use
  • Reliable
  • Reduce the risk of cross contamination and growth inhibition
The crimp cap version provides a tamperproof closure to ensure a high level of security. Manufactured in ISO® 9001 controlled environments where each lot is certifi ed for pH, sterility, and growth promotion using ATCC® strains specifi ed by the USP/EP/JP. A non-coring, large diameter septum area is easy to pierce for operator safety and productivity.