Serological pipettes, for cotton plugging
Leverancier: BELLCO GLASS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1210-02010 1210-05010 1210-10010 1210-25010 1210-50020
BELC1210-02010EA 146.69 EUR
BELC1210-02010 BELC1210-05010 BELC1210-10010 BELC1210-25010 BELC1210-50020
Serological pipettes, for cotton plugging
Pipetten Serologische pipetten
These serological pipettes are manufactured from type 1 class B borosilicate glass.
  • Constructed top for cotton plugging (supplied without cotton)
  • Calibrated to tip
  • Tempered tip; to deliver, blow-out