Racks for culture tubes, stainless steel
Leverancier: BELLCO GLASS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2027-13012 2027-13040 2027-16048 2027-16072 2027-25012 2027-25036 2027-38012 2027-38024
BELC2027-13012EA 81 EUR
BELC2027-13012 BELC2027-13040 BELC2027-16048 BELC2027-16072 BELC2027-25012 BELC2027-25036 BELC2027-38012 BELC2027-38024
Racks for culture tubes, stainless steel
Rekken Tube Racks
Made of stainless steel.
  • Very good chemical resistance
  • Very good temperature resistance
  • Sturdy
These racks are for culture tubes that will not rust or deteriorate in water baths or CO₂ atmosphere and is available in a variety of configurations for all popular tube sizes.