Colour reference solutions, ready to use, according to Ph. Eur., Certipur®, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.00265.0001 1.00266.0001 1.00267.0001 1.00268.0001 1.00269.0001
1.00265.0001EA 682 EUR
1.00265.0001 1.00266.0001 1.00267.0001 1.00268.0001 1.00269.0001
Colour reference solutions, ready to use, according to Ph. Eur., Certipur®, Supelco®
Standaarden Standaarden voor kleurreferentie en spectroscopie
Certipur® colour reference solutions are developed for ‘Method 1’ as described in chapter 2.2.2 of the European Pharmacopoeia.
  • Stable solutions with a minimum shelf life of 3 years
  • Ready to use solutions
  • Certificate of Analysis included