Microcarrier spinner flasks
Leverancier: BELLCO GLASS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1965-00100 1965-00250 1965-00500 1965-01000 1965-03000 1965-06000 1965-15000 1965-02500 1965-08000
BELC1965-00100EA 224.1 EUR
BELC1965-00100 BELC1965-00250 BELC1965-00500 BELC1965-01000 BELC1965-03000 BELC1965-06000 BELC1965-15000 BELC1965-02500 BELC1965-08000
Microcarrier spinner flasks
Kolven Spinnerflessen
Spinner flasks promotes the optimal growth of suspension and microcarrier culture.
  • Impeller height adjusts through compression fitting
  • Indented bottom (up to 1 L) prevents accumulation of microcarrier beads under impeller