Volumetric flasks, class A
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
671950 670950 672950 673950 674950 675950 676950
VITL671950EA 52.3 EUR
VITL671950 VITL670950 VITL672950 VITL673950 VITL674950 VITL675950 VITL676950
Volumetric flasks, class A
Kolven Maatkolven
PMP, opaque, with NS stoppers, PP.
  • With ring mark individually calibrated to ‘In’
  • With printed lot number and batch certificate
  • Thermal stress up to 121 °C (autoclaving) does not permanently exceed the tolerance limit
  • To preserve markings, cleaning at no higher than 60 °C is recommended
UV-absorbing, highly transparent. For storage of light-sensitive substances.
Certificaten: Class A tolerances according to DIN EN ISO 1042.