Culture shake flasks
Leverancier: BELLCO GLASS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2541-00125 2541-00250 2541-01000 2541-02000 2542-00500 2542-01000 2542-02000 2542-00250
BELC2541-00125EA 281.49 EUR
BELC2541-00125 BELC2541-00250 BELC2541-01000 BELC2541-02000 BELC2542-00500 BELC2542-01000 BELC2542-02000 BELC2542-00250
Culture shake flasks
Kolven Cultuurflessen
Flasks, made of type 1 class A borosilicate glass, with rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation.
  • With break resistant lip to resist chipping
  • Very good chemical resistance
  • Very good temperature resistance
Designed for use with orbital shakers.