Fernbach culture flasks
Leverancier: BELLCO GLASS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2551-02800 2552-62280 2552-01800 2552-02800
BELC2551-02800EA 0 EUR
BELC2551-02800 BELC2552-62280 BELC2552-01800 BELC2552-02800
Fernbach culture flasks
Kolven Cultuurflessen
These fernbach culture flasks are made of borosilicate glass type 1, class A.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These have three sided baffles with break resistant lip or triple baffled, with 45 mm PP screw cap and pouring ring. For cultures requiring a large surface area to volume ratio.