Culture flasks, DeLong®
Leverancier: BELLCO GLASS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2510-00125 2510-00250 2510-00300 2510-02000 2510-04000 2510-01000 2511-02000 2510-00500
BELC2510-00125EA 189.79 EUR
BELC2510-00125 BELC2510-00250 BELC2510-00300 BELC2510-02000 BELC2510-04000 BELC2510-01000 BELC2511-02000 BELC2510-00500
Culture flasks, DeLong®
Kolven Cultuurflessen
These DeLong® culture flasks are made of borosilicate glass type 1, class A.
  • Ideal for growing, storing and mixing of aerobic and anaerobic cultures
  • Graduated to approximate volumes
  • Very good chemical and temperature resistance
Bestelinformatie: The stainless steel or KAP-UTS™ disposable closures must be purchased separately for the flasks without closures.