Caps for culture tubes and flasks, stainless steel
Leverancier: BELLCO GLASS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2005-00013 2005-00016 2005-00018 2005-00025 2005-00038 2005-03812 2005-00020 2005-03806
BELC2005-00013EA 355.23 EUR
BELC2005-00013 BELC2005-00016 BELC2005-00018 BELC2005-00025 BELC2005-00038 BELC2005-03812 BELC2005-00020 BELC2005-03806
Caps for culture tubes and flasks, stainless steel
Sluitingen Reageerbuissluitingen
Made of type 305 stainless steel, these closures are suitable for use with culture tubes and flasks.
The fingers grip outside of the tube firmly and will not lose their resilience through repeated autoclaving or dry heat sterilization.