Magnetic stirring bars, double-ended
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0331 0332
SEMA0331EA 49.7 EUR
SEMA0331 SEMA0332
Magnetic stirring bars, double-ended
Roerders Magnetische roerstaven
Double ended magnetic stirring bars provide excellent centering, limited contact surface and high turbulence even at low speeds.
  • Ideal for test tubes or for eccentric stirring movements