Boxes, with Hinged Lid
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0341 0342 0348
SEMA0341EA 33 EUR
SEMA0341 SEMA0342 SEMA0348
Boxes, with Hinged Lid
Dozen Opbergdozen
The practical hinged flip-top box container is made from dimensionally stable Polypropylene. The standard colour is grey and the range includes a total of five different sizes. With lid and container made from a single block, snap-on cap and integral hinge, you can package your small item safely and securely.
  • Heavyweight boxes
  • Rigid and robust
  • Stackable
  • PP, grey