Tubing connectors, Y-shaped, POM
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0490 0388
SEMA0490EA 8 EUR
SEMA0490 SEMA0388
Tubing connectors, Y-shaped, POM
Fittingen en connectoren
Connectors, Y-shaped, made of POM, white.
  • Temperature resistant up to 100 °C
  • Sturdy
  • Good chemical resistance