Tubing connectors, T-shaped
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0469 0467 0392
SEMA0469EA 7.1 EUR
SEMA0469 SEMA0467 SEMA0392
Tubing connectors, T-shaped
Fittingen en connectoren
PP, translucent.
  • Good mechanical and chemical resistance
  • Very good thermal resistance
  • Autoclavable
Dimensions are shown as both nominal and exact as elasticity of tubing will influence the choice of size. Use our tubing selector 229-0111 to determine "grip". This will then give equivalent crest dimension for the particular connector concerned.