Tubing connectors, straight, reducing, PP
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0564 0563
SEMA0564EA 11.4 EUR
SEMA0564 SEMA0563
Tubing connectors, straight, reducing, PP
Fittingen en connectoren
Connectors, straight, conical shape and stepped, made of PP, wiith different fitting diameters on the two sides able to connect with tubing having different diameters.
  • Good mechanical strength and chemical resistance
  • Very good thermal resistance
  • Autoclavable