Burette clamps, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CWH002 CWH004
241-0367EA 41.8 EUR
241-0367 241-0399
Burette clamps, Azlon®
Klemmen Klemmen voor algemeen gebruik
PP, white. These spring-loaded PP clamps are corrosion resistant and can either hold one or two burettes.
  • Allow clear view of graduations along the full length of the burette
  • Rubber grips prevent slipping
  • Interchangeable compression cone on the central boss provides a firm grip on rods with Ø 11 to 14 mm