Resin, PL-AMS, StratoSpheres™
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PL1464-4689 PL1464-6689 PL1464-6749 PL1464-6769 PL1464-6789 PL1464-6799
VARIPL1464-4689EA 0 EUR
VARIPL1464-4689 VARIPL1464-6689 VARIPL1464-6749 VARIPL1464-6769 VARIPL1464-6789 VARIPL1464-6799
Resin, PL-AMS, StratoSpheres™
Sorbenten voor chromatografie Sorbenten voor SPE
Aminomethylstyrene resin is a particularly versatile material suitable for the attachment of a variety of spacers, handles and linkers (for use in solid phase synthesis).
Copolymerised PL-CMS to be used as a starting material, therefore possible for a very wide range of loadings and particle size combinations to be prepared to suit any given application. The preferred method of attachment of linkers or handles is through an amide bond.