Resin, PL-CMS, StratoSpheres™
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PL1461-4799 PL1461-6749 PL1461-6769 PL1461-6799
VARIPL1461-4799EA 0 EUR
VARIPL1461-4799 VARIPL1461-6749 VARIPL1461-6769 VARIPL1461-6799
Resin, PL-CMS, StratoSpheres™
Sorbenten voor chromatografie Sorbenten voor SPE
Chloromethylpolystyrene known as merrifield resin, PL-CMS is a copolymer support for solid phase synthesis of peptides using Boc chemistry.
Boc-amino acids are typically attached to the resin as a cesium salt, although other techniques have also been used. A slight excess of acid is neutralized with cesium carbonate and the activated acid isolated by evaporation.
Other useful techniques for cleavage include saponification or hydrolysis to create free acids, trans-esterification to create methyl esters, or aminolysis to form carboxamides. PL-CMS can be used to generate a variety of other supports by the attachment of appropriate linkers, particularly through Williamson ether synthesis.