Funnels, with short stem, lightweight, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FWC056 FWC058 FWC060 FWC062 FWC064 FWC066
AZLOFWC056EA 24.3 EUR
AZLOFWC056 221-0275 221-0276 221-0277 AZLOFWC064 AZLOFWC066
Funnels, with short stem, lightweight, Azlon®
Trechters Filtertrechters Filtertrechters
Made of PP, these translucent lightweight funnels with ribs prevent air locks, allowing rapid filtration.
  • Autoclavable to 121 °C
  • Good chemical resistance
  • Made from food grade plastic