S-Pak® Membrane Filters
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HABG047S6 HAWG047S6 HAGG050S6 HABG050S6 HCWG047S6
MILFHABG047S6EA 0 EUR
MILFHABG047S6 MILFHAWG047S6 MILXHAGG050S6 MILFHABG050S6 MILXHCWG047S6
S-Pak® Membrane Filters
Filters Membraanfilters
S-Pak™ hydrophilic sterile membranes are protected with a blue sheet of paper that keeps the membrane encountering with the film by ensuring maximum productivity and efficiency. This protective sheet prevents the migration of any adhesive.
  • Ready to use
  • Gridded filter surface
  • No hydrophobic spots and no toxicity
Used in microbiological analysis of beverages, beer, bottled water, cider, clinical lab, cosmetics, environmental analysis, food and beverage, industrial QC, soft drink, sport drink, wine and pharmaceutical products analysis.
Verpakking: Individually packed.