Erlenmeyer flasks, wide neck, with screw cap, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FDE546 FDE544
214-0112EA 32.6 EUR
214-0112 AZLOFDE544
Erlenmeyer flasks, wide neck, with screw cap, Azlon®
Kolven Erlenmeyers
PP, translucent.
  • Very good chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • Also sealable with standard ground joint stoppers (stoppers not supplied)
With blue, easy to read graduations. Autoclavable. The flasks feature a stopper bed and can also be sealed with standard ground joint polypropylene stoppers.