Powder funnels, PP, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FWC206 FWC208 FWC210 FWC212
AZLOFWC206EA 63 EUR
AZLOFWC206 221-0278 AZLOFWC210 AZLOFWC212
Powder funnels, PP, Azlon®
Trechters Poedertrechters
Funnels, made of PP, with parallel stem minimises 'bridging' of powder to improve flow.
  • Lightweight and unbreakable
  • Two largest sizes have vertical sided tops
  • Good chemical and temperature resistance