Jerricans, narrow mouth, HDPE, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JGH140 JGH148
AZLOJGH140EA 15.1 EUR
AZLOJGH140 AZLOJGH148
Jerricans, narrow mouth, HDPE, Azlon®
Flessen Jerrycans
Plastic, translucent.
  • Lightweight and robust
  • Corrosion resistant
  • Ideal for storage and transportation of liquid chemicals
  • Suitable for food contact applications in accordance with EU and U.S. FDA regulations