N-Phenylglycine ethyl ester ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
N-Phenylglycine Ethyl Ester, Phenylamino-acetic acid ethyl ester
P1781-25G
P1781-5G
TCIAP1781-25GEA
219
EUR
TCIAP1781-25G
TCIAP1781-5G
N-Phenylglycine ethyl ester ≥98.0% (by GC)
N-Phenylglycine ethyl ester
Formule:
C₁₀H₁₃NO₂ Kookpunt: 140°C |
MDL Nummer:
MFCD00038315 CAS nummer: 2216-92-4 EINECS: 218-702-2 |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 53.0 to 57.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...