N-Phenylglycine potassium salt ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Potassium 2-(phenylamino)acetate
P0179-25G
P0179-500G
TCIAP0179-25GEA
58.3
EUR
TCIAP0179-25G
TCIAP0179-500G
N-Phenylglycine potassium salt ≥98.0% (by HPLC, titration analysis)
N-Phenylglycine potassium salt
Formule:
C₈H₈KNO₂ MW: 189.25 g/mol Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00058994 CAS nummer: 19525-59-8 EINECS: 243-133-1 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...