VWR®, Tris buffer solution, pH 8 (1 mol/L) for biotechnology
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E199-100MLEA 65.2 EUR
E199-100ML E199-500ML
VWR®, Tris buffer solution, pH 8 (1 mol/L) for biotechnology
Buffers Kloonbuffers
Tris (hydroxymethyl) aminomethane.
  • Biotechnology grade
  • pH @25 °C: 7,9 to 8,1
Storage: room temperature