4-(Benzyloxy)benzaldehyde on polystyrene, cross-linked with 2% DVB 200-400 mesh
Leverancier: Novabiochem (Part of Merck)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8.55026.0100 8.55026.0025 8.55026.0005
01-64-0182-100EA 1090 EUR
01-64-0182-100 01-64-0182-25 01-64-0182-5
4-(Benzyloxy)benzaldehyde on polystyrene, cross-linked with 2% DVB 200-400 mesh
4-(Benzyloxy)benzaldehyde
Formule: C₁₄H₁₂O₂
MDL Nummer: MFCD00003387
CAS nummer: 4397-53-9