N(α)-Boc-L-histidine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Boc-His-OH

4000667.0005 4000667.0025 4000667.0100
BACE4000667.0005EA 0 EUR
BACE4000667.0005 BACE4000667.0025 BACE4000667.0100
N(α)-Boc-L-histidine
N(α)-Boc-L-histidine
Formule: C₁₁H₁₇N₃O₄
MW: 255.3 g/mol
MDL Nummer: MFCD00065576
CAS nummer: 17791-52-5
EINECS: 241-768-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR