Bis(pinacolato)diboron ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane

B1964-1G B1964-25G B1964-5G B1964-100G
TCIAB1964-1GEA 17.6 EUR
TCIAB1964-1G TCIAB1964-25G TCIAB1964-5G TCIAB1964-100G
Bis(pinacolato)diboron ≥99.0% (by GC)
Bis(pinacolato)diboron
Formule: C₁₂H₂₄B₂O₄
MDL Nummer: MFCD00799570
CAS nummer: 73183-34-3
Merck Index: 14,01300
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adea3dd4ce11ed26ddd715d93151dfc4ca405d9c&vid=adea3dd4ce11ed26ddd715d93151dfc4ca405d9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 135.0 to 142.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR