2,3-Diphenylmaleic anhydride ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
D3943-1G
D3943-5G
TCIAD3943-1GEA
148
EUR
TCIAD3943-1G
TCIAD3943-5G
2,3-Diphenylmaleic anhydride ≥95.0% (by GC)
2,3-Diphenylmaleic anhydride
Formule:
C₁₆H₁₀O₃ Kookpunt: 236°C Smeltpunt: 160°C |
MDL Nummer:
MFCD00005521 CAS nummer: 4808-48-4 EINECS: 225-370-2 |
|
Specification Test Results
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization back titration) | min. 93.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...