1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
44543.04
44543.09
44543.04EA
74
EUR
44543.04
44543.09
1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane ≥97%
1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane
Purity: 97%
CAS No.: 102488-47-1
Molecular Formula: C10H10ClF13Si
Formula Weight: 440.71
Flash Point: 52°(180°F)
Physical Form: Liquid
Density: 1.461
Refractive Index: 1.3453
Sensitivity: Moisture Sensitive
Appearance: Colourless
Boiling Point: 189-191°
Beilstein Reference: 7040835
UN No.: 2986
MDL No.: MFCD00042359
Formule:
C₁₀H₁₀ClF₁₃Si Kookpunt: 189…191 °C Dichtheid: 1,461 Vlampunt: 52 °C (180 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00042359 CAS nummer: 102488-47-1 ADR: 8,II |