10,12-Heptacosadiynoic acid ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H0666-100MG
H0666-500MG
TCIAH0666-100MGEA
102
EUR
TCIAH0666-100MG
TCIAH0666-500MG
10,12-Heptacosadiynoic acid ≥98.0% (by GC)
10,12-Heptacosadiynoic acid
Formule:
C₂₇H₄₆O₂ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00060065 CAS nummer: 67071-94-7 |
|
Specification Test Results
Appearance | White to Gray to Dark blue powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 68.0 to 73.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...