10,12-Pentacosadiynoic acid ≥97.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
P1030-1G
P1030-100MG
TCIAP1030-1GEA
70.4
EUR
TCIAP1030-1G
TCIAP1030-100MG
10,12-Pentacosadiynoic acid ≥97.0% (by GC, titration analysis)
10,12-Pentacosadiynoic acid
Formule:
C₂₅H₄₂O₂ |
MDL Nummer:
MFCD00041684 CAS nummer: 66990-32-7 |
|
Specification Test Results
Appearance | White to Gray to Dark blue powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 62.0 to 65.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...