Arsenic standard dissolved in oil, 1,000 µg/g As in mineral oil for wear metal analysis, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Specpure®
43864.1YEA 0 EUR
43864.1Y 43864.18
Arsenic standard dissolved in oil, 1,000 µg/g As in mineral oil for wear metal analysis, Specpure®
Standaarden Kationische standaarden Arseen standaarden
Physical Form: Liquid
MDL No.: MFCD00085309