Cadmium, oil based standard solutions, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42436.1Y 14228.18 42436.18
42436.1YEA 81.7 EUR
42436.1Y 14228.18 42436.18
Cadmium, oil based standard solutions, Specpure®
Standaarden Kationische standaarden Cadmium standaarden
Physical Form: Liquid
MDL No.: MFCD00010914