4-Bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylic acid
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SPB02757.250
SPB02757.1
MAYBSPB02757.250EA
0
EUR
MAYBSPB02757.250
MAYBSPB02757.1
4-Bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylic acid
4-Bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylic acid
Formule:
C₇H₉BrN₂O₂ |
MDL Nummer:
MFCD00173776 CAS nummer: 175276-99-0 |