Magnesium standard dissolved in oil, 1000 µg/g Mg in mineral oil for wear metal analysis, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14219.1Y
14219.18
14219.1YEA
0
EUR
14219.1Y
14219.18
Magnesium standard dissolved in oil, 1000 µg/g Mg in mineral oil for wear metal analysis, Specpure®
Standaarden
Kationische standaarden
Magnesium standaarden
Physical Form: Liquid
MDL No.: MFCD00085308