1-Naphthylacetamide ≥98.0% (by total nitrogen basis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
N0624-25G
N0624-500G
TCIAN0624-25GEA
27.5
EUR
TCIAN0624-25G
TCIAN0624-500G
1-Naphthylacetamide ≥98.0% (by total nitrogen basis)
1-Naphthylacetamide
Formule:
C₁₂H₁₁NO |
MDL Nummer:
MFCD00004047 CAS nummer: 86-86-2 EINECS: 201-704-2 |
|
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 182.0 to 186.0 deg-C |
Solubility in N,N-DMF | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...