Bismuth standard dissolved in oil, 5000 µg/g Bi in mineral oil for wear metal analysis, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Specpure®
43865.18EA 433 EUR
43865.18 43865.1Y
Bismuth standard dissolved in oil, 5000 µg/g Bi in mineral oil for wear metal analysis, Specpure®
Standaarden Kationische standaarden Bismut standaarden
Physical form: liquid
MDL No.: MFCD00134033