Titanium standard dissolved in oil, 1,000 µg/g Ti in mineral oil for wear metal analysis, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14204.1Y
14204.18
14204.1YEA
0
EUR
14204.1Y
14204.18
Titanium standard dissolved in oil, 1,000 µg/g Ti in mineral oil for wear metal analysis, Specpure®
Standaarden
Kationische standaarden
Titanium standaarden
Physical Form: Liquid
MDL No.: MFCD00011264